5-Fluoro-2-iodobenzoic acid 98%

Supplier: Ambeed
Danger

A145566-10G A145566-250MG A145566-1G A145566-25G A145566-5G
103803-556EA 6 USD
103803-556 103841-640 76972-820 76972-822 77316-946 77316-948 77316-950
5-Fluoro-2-iodobenzoic acid 98%
5-Fluoro-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Boiling Pt: 307.9 °C (760 mm Hg)
Melting Pt: 145…149 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00837316
CAS Number: 52548-63-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR