5-Fluorosalicylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Fluoro-2-hydroxybenzoic acid
A148218-100G
A148218-10G
A148218-1G
A148218-25G
A148218-5G
A148218-500G
103803-892EA
5
USD
103803-892
103842-006
76768-058
76768-060
77317-436
77317-438
77317-440
77597-536
5-Fluorosalicylic acid 98%
5-Fluorosalicylic acid
Precaución: for Research Use Only
Formula:
C₇H₅FO₃ Boiling Pt: 307.4 °C (760 mm Hg) Melting Pt: 177…179 °C Flash Pt: 139.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002456 CAS Number: 345-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...