5-Fluorosalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-hydroxybenzaldehyde

A214923-100G A214923-10G A214923-500G A214923-1G A214923-25G A214923-5G
103818-444EA 9 USD
103818-444 103849-144 76770-656 76770-658 76770-660 77303-386 77303-388 77303-390
5-Fluorosalicylaldehyde 98%
5-Fluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 207.4 °C (760 mm Hg)
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD01090997
CAS Number: 347-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR