5-Fluoro-2-hydrazinylpyridine dihydrochloride 95%

Supplier: Ambeed
Warning

A153100-100MG A153100-10G A153100-250MG
103804-464EA 38 USD
103804-464 103842-634 76984-716 76984-718 76984-720
5-Fluoro-2-hydrazinylpyridine dihydrochloride 95%
5-Fluoro-2-hydrazinylpyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₆FN₃·2ClH
MW: 200.042 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20441448
CAS Number: 1401426-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR