5-Fluoro-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-7-azaindole

A143407-100MG A143407-10G A143407-250MG A143407-25G A143407-5G
103803-274EA 19 USD
103803-274 77162-438 77162-440 77162-442 77299-336 77299-338
5-Fluoro-1H-pyrrolo[2,3-b]pyridine 97%
5-Fluoro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C7H5FN2
MW: 136.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659678
CAS Number: 866319-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR