5-Fluoro-1H-indole-3-carbonitrile 98%

Supplier: Ambeed
Warning

A130979-250MG A130979-5G
103801-510EA 46 USD
103801-510 77271-998 77298-446
5-Fluoro-1H-indole-3-carbonitrile 98%
5-Fluoro-1H-indole-3-carbonitrile

Precaución: for Research Use Only

Formula: C9H5FN2
MW: 160.1500 g/mol
Boiling Pt: 353.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06809706
CAS Number: 194490-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR