5-Fluoro-1H-indole-3-carbaldehyde 96%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoroindole-3-carboxaldehyde

A297639-1G A297639-250MG A297639-25G
103814-146EA 30 USD
103814-146 103835-288 76837-974 77277-274 77305-712
5-Fluoro-1H-indole-3-carbaldehyde 96%
5-Fluoro-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆FNO
MW: 163.15 g/mol
Boiling Pt: 342.4 °C (760 mm Hg)
Melting Pt: 164 °C
Flash Pt: 160.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00022719
CAS Number: 2338-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR