5-Fluoroindole 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-1H-indole

A170102-1G A170102-25G A170102-5G A170102-100G A170102-10G A170102-500G
103806-450EA 6 USD
103806-450 103844-716 77081-814 77081-816 77081-818 77273-668 77273-670 77273-672
5-Fluoroindole 98% (by GC)
5-Fluoroindole

Precaución: for Research Use Only

Formula: C₈H₆FN
Boiling Pt: 258 °C (760 mm Hg)
Melting Pt: 45…48 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005671
CAS Number: 399-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR