5-Fluoro-1,3-dihydrobenzoimidazol-2-one 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-1H-benzo[d]imidazol-2(3H)-one

A259644-10G A259644-250MG A259644-1G A259644-5G
103812-454EA 18 USD
103812-454 103852-336 76771-846 76771-848 77091-908 77091-910
5-Fluoro-1,3-dihydrobenzoimidazol-2-one 98%
5-Fluoro-1,3-dihydrobenzoimidazol-2-one

Precaución: for Research Use Only

Formula: C₇H₅FN₂O
MW: 152.13 g/mol
Melting Pt: 303 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663528
CAS Number: 1544-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR