5-Ethynyl-2'-deoxyuridine 98%

Supplier: Ambeed
Warning

Synonyms: 2′-Deoxy-5-ethynyluridine, 5-Ethynyl-2'-dU, EdU

A254440-250MG A254440-100MG
103812-136EA 88 USD
103812-136 103851-990 76771-764 77626-334
5-Ethynyl-2'-deoxyuridine 98%
5-Ethynyl-2'-deoxyuridine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₅
MW: 252.23 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01675687
CAS Number: 61135-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR