2-Amino-5-ethyl-pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Ethylpyridin-2-amine

A281744-1G A281744-5G A281744-250MG
103799-562EA 26 USD
103799-562 103813-568 103853-518 76837-214 76837-216 77250-510
2-Amino-5-ethyl-pyridine 95%
2-Amino-5-ethyl-pyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09757479
CAS Number: 19842-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR