5-Ethyl-4-hydroxy-2-methylfuran-3(2H)-one 97%

Supplier: Ambeed
Warning

A127174-100G A127174-10G A127174-25G A127174-5G
77108-962EA 176.14 USD
77108-962 77108-964 77298-186 77298-188
5-Ethyl-4-hydroxy-2-methylfuran-3(2H)-one 97%
5-Ethyl-4-hydroxy-2-methylfuran-3(2H)-one

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
Boiling Pt: 231 °C (760 mm Hg)
Flash Pt: 93.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03411593
CAS Number: 27538-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR