5-Ethoxy-2-pyridine carboxylic acid 97%

Supplier: Ambeed

Synonyms: 5-Ethoxypicolinic acid

A126379-250MG A126379-1G A126379-5G
103800-802EA 106 USD
103800-802 76971-890 77298-118 77298-120
5-Ethoxy-2-pyridine carboxylic acid 97%
5-Ethoxy-2-pyridine carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.17 g/mol
Boiling Pt: 321.3±22.0 °C (760 mm Hg)
MDL Number: MFCD13562922
CAS Number: 98353-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR