2-Amino-5-ethoxy-6-methylpyridine 95%
Supplier: Ambeed
|
Warning
|
A333292-100MG
A333292-250MG
103825-630EA
200
USD
103825-630
103836-596
76773-094
76773-096
2-Amino-5-ethoxy-6-methylpyridine 95%
2-Amino-5-ethoxy-6-methylpyridine
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂O Boiling Pt: 261.3 °C (760 mm Hg) Flash Pt: 111.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD10000613 CAS Number: 73101-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...