2-Amino-5-cyclopropylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Cyclopropylpyridin-2-amine

A110830-250MG A110830-1G
103797-738EA 125 USD
103797-738 103823-546 77025-762 77310-344
2-Amino-5-cyclopropylpyridine 95%
2-Amino-5-cyclopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂
MW: 134.178 g/mol
Boiling Pt: 273.5±28.0 °C (760 mm Hg)
Flash Pt: 143.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06802676
CAS Number: 893738-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR