5-Cyclopropyl-isoxazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Cyclopropylisoxazole-3-carboxylic acid

A332421-1G A332421-5G
103825-608EA 53 USD
103825-608 103836-574 76839-572 76839-574
5-Cyclopropyl-isoxazole-3-carboxylic acid 95%
5-Cyclopropyl-isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 96…100 °C
Flash Pt: 170.4 °C
Storage Temperature: Freezer
MDL Number: MFCD05668325
CAS Number: 110256-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR