5-Cyclopropyl-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

A274748-100G A274748-10G A274748-1G A274748-25G A274748-5G
103813-216EA 7 USD
103813-216 103853-144 76816-284 76816-286 77092-974 77092-976 77092-978
5-Cyclopropyl-1H-pyrazol-3-amine 95%
5-Cyclopropyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃
Boiling Pt: 367.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00067985
CAS Number: 175137-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR