5-Cyanopyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A249316-100MG A249316-250MG A249316-1G
103811-864EA 383 USD
103811-864 76987-842 76987-844 77304-478
5-Cyanopyrazine-2-carboxylic acid 97%
5-Cyanopyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃N₃O₂
Boiling Pt: 328.6±15.0 °C (760 mm Hg)
Flash Pt: 152.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22070527
CAS Number: 1211533-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR