5-cyanopicolinic acid 97%

Supplier: Ambeed
Danger

Synonyms: 5-Cyanopyridine-2-carboxylic acid

A138582-250MG A138582-1G A138582-5G A138582-10G
103802-634EA 12 USD
103802-634 76767-680 77315-674 77315-676 77624-156
5-cyanopicolinic acid 97%
5-cyanopicolinic acid

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂
MW: 148.121 g/mol
Boiling Pt: 370.7 °C (760 mm Hg)
Melting Pt: 210…215 °C
Flash Pt: 178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09152775
CAS Number: 53234-55-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR