5-Cyanoindole-3-carboxyaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3-Formyl-1H-indole-5-carbonitrile

A176749-10G A176749-1G A176749-25G A176749-100G
103807-172EA 15 USD
103807-172 103845-498 76998-394 77082-856 77082-858 77881-344
5-Cyanoindole-3-carboxyaldehyde 97%
5-Cyanoindole-3-carboxyaldehyde

Precaución: for Research Use Only

Formula: C10H6N2O
Boiling Pt: 429.6 °C (760 mm Hg)
Melting Pt: 248…253 °C
Flash Pt: 213.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01719101
CAS Number: 17380-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR