5-Cyano-2-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methylnicotinonitrile, 5-Cyano-2-picoline

A116401-10G A116401-250MG A116401-25G A116401-1G A116401-5G
103798-728EA 13 USD
103798-728 103824-592 76995-542 76995-544 76995-546 77311-402 77311-404
5-Cyano-2-methylpyridine 98%
5-Cyano-2-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 223.5 °C (760 mm Hg)
Melting Pt: 83…85 °C
Flash Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00038042
CAS Number: 3222-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR