5-Cyano-2-fluorophenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Cyano-2-fluorobenzeneboronic acid
A290754-1G
A290754-25G
A290754-5G
A290754-100MG
A290754-250MG
103813-876EA
10
USD
103813-876
103835-018
76837-602
76837-604
76837-606
77141-324
77141-326
5-Cyano-2-fluorophenylboronic acid 97%
5-Cyano-2-fluorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BFNO₂ Boiling Pt: 351.3 °C (760 mm Hg) Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095147 CAS Number: 468718-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...