5-Cyano-2-fluorobenzene-1-sulfonyl chloride 97%

Supplier: Ambeed
Danger

A361891-100MG A361891-250MG A361891-1G
103826-292EA 55 USD
103826-292 103837-306 76876-394 76876-396 77306-726
5-Cyano-2-fluorobenzene-1-sulfonyl chloride 97%
5-Cyano-2-fluorobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClFNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD18394026
CAS Number: 1101120-80-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR