5-Chlorothiophene-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A103252-100MG A103252-250MG
103796-518EA 19 USD
103796-518 103822-220 76765-290 76765-292
5-Chlorothiophene-3-carboxylic acid 97%
5-Chlorothiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C5H3ClO2S
MW: 162.5900 g/mol
Boiling Pt: 287 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00082877
CAS Number: 36157-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR