5-Chlorothiophene-2-carbonyl chloride 97%

Supplier: Ambeed
Danger

A839211-100G A839211-10G A839211-1G A839211-25G A839211-5G
103841-202EA 11 USD
103841-202 76776-680 76776-682 76776-684 76845-440 76845-442
5-Chlorothiophene-2-carbonyl chloride 97%
5-Chlorothiophene-2-carbonyl chloride

Precaución: for Research Use Only

Formula: C5H2Cl2OS
MW: 181.04 g/mol
Boiling Pt: 223.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00043886
CAS Number: 42518-98-9
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR