5-Chloropyrimidine-2-carbonitrile 95%

Supplier: Ambeed
Warning

A392644-1G A392644-5G
103826-904EA 165 USD
103826-904 103837-942 77307-012 77307-014
5-Chloropyrimidine-2-carbonitrile 95%
5-Chloropyrimidine-2-carbonitrile

Precaución: for Research Use Only

Formula: C5H2ClN3
MW: 139.5400 g/mol
Boiling Pt: 292 °C (760 mm Hg)
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD03001466
CAS Number: 38275-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR