5-Chloropyrimidine 98%

Supplier: Ambeed
Warning

A131942-10G A131942-250MG A131942-25G
103801-660EA 43 USD
103801-660 76811-232 76811-234 77298-522
5-Chloropyrimidine 98%
5-Chloropyrimidine

Precaución: for Research Use Only

Formula: C4H3ClN2
Boiling Pt: 172.9 °C (760 mm Hg)
Flash Pt: 72.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03840717
CAS Number: 17180-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR