5-Chloropyrido[3,4-b]pyrazine 97%

Supplier: Ambeed
Warning

A230364-100MG A230364-250MG A230364-5G
103819-542EA 46 USD
103819-542 103850-440 76771-124 76771-126 77089-298
5-Chloropyrido[3,4-b]pyrazine 97%
5-Chloropyrido[3,4-b]pyrazine

Precaución: for Research Use Only

Formula: C7H4ClN3
MW: 165.5800 g/mol
Boiling Pt: 314.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08706209
CAS Number: 214045-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR