5-Chloropyridine-2-carboxamide 98%

Supplier: Ambeed
Warning

A100066-10G A100066-1G
103795-230EA 26 USD
103795-230 103820-952 76765-094 77296-560
5-Chloropyridine-2-carboxamide 98%
5-Chloropyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Melting Pt: 205…209 °C
Storage Temperature: Ambient
MDL Number: MFCD10574693
CAS Number: 370104-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR