3-Amino-5-chloropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloropyridin-3-amine

A275517-10G A275517-250MG A275517-1G A275517-5G A275517-25G
103813-262EA 14 USD
103813-262 103853-186 77002-038 77002-040 77093-038 77093-040 77305-198
3-Amino-5-chloropyridine 98%
3-Amino-5-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 275.8 °C (760 mm Hg)
Flash Pt: 120.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03701386
CAS Number: 22353-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR