5-Chloropyridazine-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A207212-100MG
A207212-250MG
A207212-1G
103817-772EA
515
USD
103817-772
103848-450
76814-470
76814-472
77303-022
5-Chloropyridazine-3-carboxylic acid 95%
5-Chloropyridazine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Boiling Pt: 420.3±25.0 °C (760 mm Hg) Flash Pt: 208 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD20659173 CAS Number: 1211587-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...