5-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile 98%

Supplier: Ambeed
Warning

A280775-100MG A280775-250MG
103813-518EA 75 USD
103813-518 76988-480 76988-482
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile 98%
5-Chloropyrazolo[1,5-a]pyrimidine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClN₄
MW: 178.579 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18073765
CAS Number: 1224288-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR