5-Chloropyrazolo[1,5-a]pyridine 98%

Supplier: Ambeed
Warning

A158925-100MG A158925-250MG
103805-194EA 108 USD
103805-194 103843-406 77011-126 77011-128
5-Chloropyrazolo[1,5-a]pyridine 98%
5-Chloropyrazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21603615
CAS Number: 1101120-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR