5-Chloropyrazine-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A184178-100G
A184178-10G
A184178-25G
A184178-5G
A184178-1G
103815-722EA
6
USD
103815-722
103846-284
76769-656
76769-658
77083-896
77083-898
77301-884
5-Chloropyrazine-2-carboxylic acid 98%
5-Chloropyrazine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C5H3ClN2O2 Boiling Pt: 330.9 °C (760 mm Hg) Flash Pt: 153.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD09033269 CAS Number: 36070-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...