5-Chloronicotinic acid 97%

Supplier: Ambeed
Warning

A213623-5G A213623-100G A213623-10G A213623-250MG A213623-1G
103818-312EA 14 USD
103818-312 103849-020 77087-530 77097-782 77097-784 77097-786 77303-304
5-Chloronicotinic acid 97%
5-Chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
Boiling Pt: 303.7 °C (760 mm Hg)
Melting Pt: 169…171 °C
Flash Pt: 137.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00128901
CAS Number: 22620-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR