3-Chloro-5-formylpyridine 97%

Supplier: Ambeed
Danger

A382554-1G A382554-5G A382554-100MG A382554-250MG A382554-25G
103826-718EA 47 USD
103826-718 103837-754 76841-074 76841-076 77016-082 77016-084 77890-791
3-Chloro-5-formylpyridine 97%
3-Chloro-5-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄ClNO
Boiling Pt: 226.026 °C (760 mm Hg)
Melting Pt: 70…71 °C
Flash Pt: 90.498 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08457773
CAS Number: 113118-82-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR