5-Chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide 2-aminoethanol salt 98%

Supplier: Ambeed
Warning

A116636-100G A116636-25G
103808-158EA 84 USD
103808-158 77108-274 77108-276
5-Chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide 2-aminoethanol salt 98%
5-Chloro-N-(2-chloro-4-nitrophenyl)-2-hydroxybenzamide 2-aminoethanol salt

Precaución: for Research Use Only

Formula: C₁₅H₁₅Cl₂N₃O₅
MW: 388.2 g/mol
Boiling Pt: 424.5 °C (760 mm Hg)
Flash Pt: 210.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01941566
CAS Number: 1420-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR