5-Chloroisoindoline hydrochloride 98%

Supplier: Ambeed
Warning

A114841-1G A114841-250MG
103798-490EA 65 USD
103798-490 103824-332 77297-338 77670-608
5-Chloroisoindoline hydrochloride 98%
5-Chloroisoindoline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₈ClN·ClH
MW: 190.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11223562
CAS Number: 912999-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR