4-Chlorophthalic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloroisobenzofuran-1,3-dione

A311361-25G A311361-1000G A311361-100G A311361-10G A311361-500G A311361-5G
103835-848EA 5 USD
103835-848 76838-644 77194-652 77194-654 77194-656 77194-658 77306-050
4-Chlorophthalic anhydride 97%
4-Chlorophthalic anhydride

Precaución: for Research Use Only

Formula: C₈H₃ClO₃
MW: 182.56 g/mol
Boiling Pt: 295 °C (760 mm Hg)
Melting Pt: 96 °C
Flash Pt: 142.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00152354
CAS Number: 118-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR