5-Chloro-1H-indole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloroindole-3-carboxaldehyde

A197702-10G A197702-250MG A197702-1G A197702-25G A197702-5G
103816-958EA 14 USD
103816-958 103847-580 77030-044 77030-046 77085-596 77302-564 77302-566
5-Chloro-1H-indole-3-carbaldehyde 98%
5-Chloro-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MW: 179.61 g/mol
Melting Pt: 213…216 °C
Flash Pt: 179.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00175291
CAS Number: 827-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR