5-Chloroindole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-1H-indole

A144835-100G A144835-10G A144835-1G A144835-25G A144835-5G
103803-456EA 5 USD
103803-456 103841-534 77272-682 77272-684 77299-414 77316-788 77316-790
5-Chloroindole 98%
5-Chloroindole

Precaución: for Research Use Only

Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 293 °C (760 mm Hg)
Melting Pt: 69…71 °C
Flash Pt: 158.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005672
CAS Number: 17422-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR