5-Chloro-2-furoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chlorofuran-2-carboxylic acid

A305386-10G A305386-250MG A305386-1G A305386-25G A305386-5G
103814-430EA 17 USD
103814-430 103835-596 76772-710 76772-712 76838-294 76838-296 76838-298
5-Chloro-2-furoic acid 97%
5-Chloro-2-furoic acid

Precaución: for Research Use Only

Formula: C₅H₃ClO₃
MW: 146.529 g/mol
Flash Pt: 103.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00093272
CAS Number: 618-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR