5-Chloro-7-nitro-1H-indole 97%

Supplier: Ambeed
Warning

A106198-100MG A106198-10G A106198-250MG A106198-1G A106198-5G A106198-25G
103797-008EA 27 USD
103797-008 103822-762 77148-486 77148-488 77148-490 77309-622 77309-624 77900-695
5-Chloro-7-nitro-1H-indole 97%
5-Chloro-7-nitro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
Boiling Pt: 379.6±22.0 °C (760 mm Hg)
Flash Pt: 183.4 °C
Storage Temperature: Ambient
MDL Number: MFCD20923231
CAS Number: 1197181-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR