5-Chloro-7-methylisatin 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-7-methylindoline-2,3-dione, 5-Chloro-7-methyl-1H-indole-2,3-dione

A295065-10G A295065-250MG A295065-1G A295065-25G A295065-5G
103814-042EA 38 USD
103814-042 103835-178 76693-310 76693-312 76837-826 76837-828 76837-830
5-Chloro-7-methylisatin 97%
5-Chloro-7-methylisatin

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MW: 195.6 g/mol
Melting Pt: 283 °C
Storage Temperature: Ambient
MDL Number: MFCD00047218
CAS Number: 14389-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR