5-Chloro-6-methylpyridin-3-ol 98%

Supplier: Ambeed
Warning

A131479-100MG A131479-250MG A131479-1G
103801-586EA 413 USD
103801-586 77009-974 77009-976 77314-092
5-Chloro-6-methylpyridin-3-ol 98%
5-Chloro-6-methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610476
CAS Number: 51984-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR