Etoricoxib 99%

Supplier: Ambeed
Danger

Synonyms: MK0663, L-791456, MK-0663, 5-Chloro-6'-methyl-3-[4-(methylsulphonyl)phenyl]-2,3'-bipyridine, Arcoxia, 5-Chloro-6'-methyl-3-(4-(methylsulphonyl)phenyl)-2,3'-bipyridine

A137375-100MG A137375-10MG A137375-50MG A137375-1G A137375-250MG A137375-1MG
103802-448EA 64 USD
103802-448 76798-104 76798-106 76798-108 77298-904 77566-744 77666-982
Etoricoxib 99%
Etoricoxib

Precaución: For research use only.

Formula: C₁₈H₁₅ClN₂O₂S
MW: 358.84 g/mol
Boiling Pt: 510 °C (760 mm Hg)
Melting Pt: 134…135 °C
Flash Pt: 262.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797512
CAS Number: 202409-33-4
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR