5-Amino-3-chloro-2-methoxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-6-methoxypyridin-3-amine

A134052-250MG A134052-25G A134052-1G A134052-5G
103801-964EA 31 USD
103801-964 76983-882 77298-686 77314-674 77314-676
5-Amino-3-chloro-2-methoxypyridine 97%
5-Amino-3-chloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.586 g/mol
Boiling Pt: 255.4 °C (760 mm Hg)
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD13190367
CAS Number: 158387-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR