5-Chloro-6-iodo-2-(methylsulfonyl)-1H-benzo[d]imidazole 97%
Supplier: Ambeed
|
Warning
|
A130101-100MG
A130101-250MG
A130101-1G
103801-378EA
88
USD
103801-378
77009-878
77009-880
77298-388
5-Chloro-6-iodo-2-(methylsulfonyl)-1H-benzo[d]imidazole 97%
5-Chloro-6-iodo-2-(methylsulfonyl)-1H-benzo[d]imidazole
Precaución: for Research Use Only
Formula:
C₈H₆ClIN₂O₂S MW: 356.57 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24387215 CAS Number: 1219741-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...