5-Chloro-6-fluoropyridin-3-ol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-fluoro-5-hydroxypyridine

A235649-100MG A235649-250MG A235649-25G A235649-1G A235649-5G
103811-024EA 46 USD
103811-024 103850-792 77000-298 77000-300 77000-302 77089-802 77089-804
5-Chloro-6-fluoropyridin-3-ol 97%
5-Chloro-6-fluoropyridin-3-ol

Precaución: for Research Use Only

Formula: C5H3ClFNO
MW: 147.53 g/mol
Boiling Pt: 354.4 °C (760 mm Hg)
Flash Pt: 168.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08061311
CAS Number: 209328-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR