Methyl 5-chloro-4-nitro-thiophene-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A503245-1G
A503245-5G
A503245-10G
A503245-250MG
103828-310EA
24
USD
103828-310
103839-404
76843-374
76843-376
77103-022
77103-024
Methyl 5-chloro-4-nitro-thiophene-2-carboxylate 95%
Methyl 5-chloro-4-nitro-thiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₄ClNO₄S Boiling Pt: 358.2 °C (760 mm Hg) Melting Pt: 80…84 °C Flash Pt: 170.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08275718 CAS Number: 57800-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...